missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin Q-Acid 98.0+%, TCI America™

Catalog No. L02381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02381G 1 g
L02385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02381G Supplier TCI America Supplier No. L02381G
Only null left

Chemical Identifiers

CAS 100986-89-8
Molecular Formula C13H9F2NO4
Molecular Weight (g/mol) 281.215
MDL Number MFCD04039905
InChI Key NVKWWNNJFKZNJO-YFKPBYRVSA-N
Synonym levofloxacin q-acid, levofloxacin carboxylic acid, s-9,10-difluoro-3-methyl-7-oxo-3,7-dihydro-2h-1,4 oxazino 2,3,4-ij quinoline-6-carboxylic acid, unii-08gt8fy84e, levofloxacin related compound b, levofloxacin acid, 3s-9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, levofloxacin q-acid,-, s-9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, levofloxacin related compound b usp
PubChem CID 688333
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 281.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Levofloxacin Q-Acid