accessibility menu, dialog, popup

UserName

H3CONOFFOOH

CAS RN 100986-89-8

IUPAC Name: 6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Synonyms: 6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Molecular Weight (g/mol): 281.22
Molecular Formula: C13H9F2NO4
InChi Key: NVKWWNNJFKZNJO-UHFFFAOYNA-N
SMILES: CC1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2F
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)