missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levofloxacin Hemihydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 138199-71-0 |
| Linear Formula | C18H20 FN3O4 · 0.5H2 O |
| MDL Number | MFCD07772024 |
| Synonym | Levofloxacin hemihydrate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 370.38 |
| Grade | Certified Reference Material |