missing translation for 'onlineSavingsMsg'
Learn More

Levocetirizine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7131899
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02641G 1 g
L02645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02641G Supplier TCI America Supplier No. L02641G

Chemical Identifiers

CAS 130018-87-0
Molecular Formula C21H27Cl3N2O3
Molecular Weight (g/mol) 461.808
MDL Number MFCD07366507
InChI Key PGLIUCLTXOYQMV-GHVWMZMZSA-N
Synonym 2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride
PubChem CID 9955977
IUPAC Name 2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
SMILES C1CN(CCN1CCOCC(=O)O)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl.Cl.Cl

Specifications

Melting Point 214°C
Color White
Quantity 1 g
Formula Weight 461.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Levocetirizine Dihydrochloride
RTECSNumber AG0990000
TSCA No