missing translation for 'onlineSavingsMsg'
Learn More

Levitiracetam Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012327
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012327 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012327 Supplier Merck Emd Millipore Supplier No. PHR14471G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Levitiracetam, an analog of piracetam, is an antiepileptic drug widely used for the remedy of epileptic seizures, partial as well as generalized epilepsy.

Specifications

CAS 102767-28-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H14N2O2
MDL Number MFCD03265610
Synonym Levetiracetam; (alphaS)-alpha-Ethyl-2-oxo-1-pyrrolidineacetamide; 2(S)-(2-Oxopyrrolidin-1-yl)butyramide
Type Certified Reference Material
Quantity 1 g
Formula Weight 170.21
Grade Certified Reference Material