missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 462120250

 View more versions of this product

Catalog No. AC462120250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Levetiracetam
C8H14N2O2
25g
levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
HPHUVLMMVZITSG-ZCFIWIBFSA-N
(2R)-2-(2-oxopyrrolidin-1-yl)butanamide
5284583
170.21
102767-28-2
MFCD03265610
-76
Solubility in water: 104 g/100 mL. Other solubilities: soluble in chloroform, methanol, acetone
CC[C@@H](N1CCCC1=O)C(N)=O
170.21
CHEBI:6437

Chemical Identifiers

102767-28-2
170.21
HPHUVLMMVZITSG-ZCFIWIBFSA-N
5284583
(2R)-2-(2-oxopyrrolidin-1-yl)butanamide
C8H14N2O2
MFCD03265610
levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
CHEBI:6437
CC[C@@H](N1CCCC1=O)C(N)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only