missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levetiracetam, Thermo Scientific™
$168.27 - $511.28
Chemical Identifiers
| CAS | 102767-28-2 |
|---|---|
| Molecular Formula | C8H14N2O2 |
| Molecular Weight (g/mol) | 170.21 |
| MDL Number | MFCD03265610 |
| InChI Key | HPHUVLMMVZITSG-ZCFIWIBFSA-N |
| Synonym | levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa |
| PubChem CID | 5284583 |
| ChEBI | CHEBI:6437 |
| IUPAC Name | (2R)-2-(2-oxopyrrolidin-1-yl)butanamide |
| SMILES | CC[C@@H](N1CCCC1=O)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC462120050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC462120250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 102767-28-2 | |
| 170.21 | |
| HPHUVLMMVZITSG-ZCFIWIBFSA-N | |
| 5284583 | |
| (2R)-2-(2-oxopyrrolidin-1-yl)butanamide |
| C8H14N2O2 | |
| MFCD03265610 | |
| levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa | |
| CHEBI:6437 | |
| CC[C@@H](N1CCCC1=O)C(N)=O |
Specifications
| 102767-28-2 | |
| MFCD03265610 | |
| levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa | |
| HPHUVLMMVZITSG-ZCFIWIBFSA-N | |
| (2R)-2-(2-oxopyrrolidin-1-yl)butanamide | |
| 5284583 | |
| 170.21 |
| C8H14N2O2 | |
| -76 | |
| Solubility in water: 104 g/100 mL. Other solubilities: soluble in chloroform, methanol, acetone | |
| CC[C@@H](N1CCCC1=O)C(N)=O | |
| 170.21 | |
| CHEBI:6437 | |
| Levetiracetam |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only