missing translation for 'onlineSavingsMsg'
Learn More

Levamisole Hydrochloride 99.0+%, TCI America™

Catalog No. p-7136661
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T121510G 10 g
T121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T121510G Supplier TCI America Supplier No. T121510G

Chemical Identifiers

CAS 16595-80-5
Molecular Formula C11H13ClN2S
Molecular Weight (g/mol) 240.749
MDL Number MFCD00012675
InChI Key LAZPBGZRMVRFKY-HNCPQSOCSA-N
Synonym levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
PubChem CID 27944
ChEBI CHEBI:6433
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
SMILES C1CSC2=NC(CN21)C3=CC=CC=C3.Cl

Specifications

Melting Point 230°C
Color White
Quantity 10 g
UN Number 2811
Formula Weight 240.75
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Levamisole Hydrochloride
RTECSNumber NJ5960000
TSCA No