missing translation for 'onlineSavingsMsg'
Learn More

Levamisole Hydrochloride 99.0+%, TCI America™

Catalog No. p-7131880
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02311G 1 g
L02315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02311G Supplier TCI America Supplier No. L02311G
Only null left

Chemical Identifiers

CAS 16595-80-5
Molecular Formula C11H13ClN2S
Molecular Weight (g/mol) 240.749
MDL Number MFCD00012675
InChI Key LAZPBGZRMVRFKY-HNCPQSOCSA-N
Synonym levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
PubChem CID 27944
ChEBI CHEBI:6433
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
SMILES C1CSC2=NC(CN21)C3=CC=CC=C3.Cl

Specifications

Melting Point 230°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 240.75
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Levamisole Hydrochloride [for Biochemical Research]
RTECSNumber NJ5960000
TSCA No