missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levamisole hydrochloride, 99.62%, MP Biomedicals™
$121.51 - $121.51
Chemical Identifiers
| CAS | 16595-80-5 |
|---|---|
| Molecular Formula | C11H13ClN2S |
| Molecular Weight (g/mol) | 240.749 |
| InChI Key | LAZPBGZRMVRFKY-HNCPQSOCSA-N |
| Synonym | levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris |
| PubChem CID | 27944 |
| ChEBI | CHEBI:6433 |
| IUPAC Name | (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride |
| SMILES | C1CSC2=NC(CN21)C3=CC=CC=C3.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15522810
|
MP Biomedicals Inc
0215522810 |
10 g |
Each for $121.51
|
|
|||||
Chemical Identifiers
| 16595-80-5 | |
| 240.749 | |
| levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris | |
| CHEBI:6433 | |
| C1CSC2=NC(CN21)C3=CC=CC=C3.Cl |
| C11H13ClN2S | |
| LAZPBGZRMVRFKY-HNCPQSOCSA-N | |
| 27944 | |
| (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride |
Specifications
| 16595-80-5 | |
| C11H13ClN2S | |
| levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris | |
| C1CSC2=NC(CN21)C3=CC=CC=C3.Cl | |
| 240.749 | |
| CHEBI:6433 | |
| 0.9962 | |
| Levamisole hydrochloride |
| White | |
| 10 g | |
| LAZPBGZRMVRFKY-HNCPQSOCSA-N | |
| (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride | |
| 27944 | |
| 240.8 | |
| Crystalline Powder |