missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Levamisole hydrochloride, 99+%

Supplier:  Thermo Scientific™ 187870500

Catalog No. AC187870500


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Levamisole hydrochloride
0.5% max. (1 g, 105°C, 4 hrs)
99+%
C11H12N2S·ClH
MFCD00012675
− 127.50 (20.00°C c=5,H2O)
levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
LAZPBGZRMVRFKY-HNCPQSOCSA-N
(6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
27944
240.76
16595-80-5
Authentic
Glass bottle
50g
15, 5511
-127.5
Solubility in water: 210g/L (20°C). Other solubilities: soluble in methanol and propylene glycol,sparingly soluble in ethanol,slightly soluble in chloroform,hexane and acetone
C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
240.76
CHEBI:6433
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 240-654-6

SDS
Documents

Documents

Promotions

Promotions