missing translation for 'onlineSavingsMsg'
Learn More

Leupeptin Trifluoroacetate Salt, MP Biomedicals

Catalog No. p-3178986
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19562301 1 mg
ICN19562305 5 mg
ICN19562325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19562301 Supplier MP Biomedicals Inc Supplier No. 0219562301

Chemical Identifiers

CAS 147385-61-3
Molecular Formula C22H39F3N6O6
Molecular Weight (g/mol) 540.59
MDL Number MFCD00133458
InChI Key WUHGBZVQELGOMH-UHFFFAOYNA-N
Synonym leupeptin trifluoroacetate salt, n-5-carbamimidamido-1-oxopentan-2-yl-2-2-acetamido-4-methylpentanamido-4-methylpentanamide; trifluoroacetic acid, trifluoroacetic acid-n-acetylleucyl-n-5-diaminomethylidene amino-1-oxopentan-2-yl leucinamide 1/1
PubChem CID 44134799
IUPAC Name N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide; trifluoroacetic acid
SMILES OC(=O)C(F)(F)F.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O

Specifications

Quantity 1 mg
Formula Weight 540.6
Chemical Name or Material Leupeptin Trifluoroacetate Salt