accessibility menu, dialog, popup

UserName

H3CCH3ONHH3CH3COHNNH2NH2NOHNOCH3OFFFHO

CAS RN 147385-61-3

IUPAC Name: N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide; trifluoroacetic acid
Synonyms: N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide; trifluoroacetic acid
Molecular Weight (g/mol): 540.59
Molecular Formula: C22H39F3N6O6
InChi Key: WUHGBZVQELGOMH-UHFFFAOYNA-N
SMILES: OC(=O)C(F)(F)F.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)