missing translation for 'onlineSavingsMsg'
Learn More

Leupeptin Hydrochloride, MP Biomedicals

$80.58 - $1,517.93

Chemical Identifiers

CAS 24125-16-4
Molecular Formula C20H39ClN6O4
Molecular Weight (g/mol) 463.02
InChI Key OQQYPQNFLLAILR-UHFFFAOYSA-N
Synonym leupeptin hydrochloride, n-5-carbamimidamido-1-oxopentan-2-yl-2-2-acetamido-4-methylpentanamido-4-methylpentanamide hydrochloride, acetyl-leu-leu-arg-al nieani, leupeptin hydrochloride, microbial hplc, n-acetylleucyl-n-5-diaminomethylidene amino-1-oxopentan-2-yl leucinamide-hydrogen chloride 1/1
PubChem CID 16219551
IUPAC Name 2-acetamido-N-[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]-4-methylpentanamide;hydrochloride
SMILES CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O)NC(=O)C.Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19562401
View Documents
MP Biomedicals Inc
0219562401
1 mg N/A
Only null left
 
ICN19562405
View Documents
MP Biomedicals Inc
0219562405
5 mg N/A
Only null left
 
ICN19562425
View Documents
MP Biomedicals Inc
0219562425
25 mg N/A
Only null left
 
ICN19562480
View Documents
MP Biomedicals Inc
0219562480
100 mg N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

24125-16-4
463.02
leupeptin hydrochloride, n-5-carbamimidamido-1-oxopentan-2-yl-2-2-acetamido-4-methylpentanamido-4-methylpentanamide hydrochloride, acetyl-leu-leu-arg-al nieani, leupeptin hydrochloride, microbial hplc, n-acetylleucyl-n-5-diaminomethylidene amino-1-oxopentan-2-yl leucinamide-hydrogen chloride 1/1
2-acetamido-N-[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]-4-methylpentanamide;hydrochloride
C20H39ClN6O4
OQQYPQNFLLAILR-UHFFFAOYSA-N
16219551
CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O)NC(=O)C.Cl

Specifications

24125-16-4
1 mg
OQQYPQNFLLAILR-UHFFFAOYSA-N
2-acetamido-N-[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]-4-methylpentanamide;hydrochloride
16219551
Leupeptin Hydrochloride
C20H39ClN6O4
leupeptin hydrochloride, n-5-carbamimidamido-1-oxopentan-2-yl-2-2-acetamido-4-methylpentanamido-4-methylpentanamide hydrochloride, acetyl-leu-leu-arg-al nieani, leupeptin hydrochloride, microbial hplc, n-acetylleucyl-n-5-diaminomethylidene amino-1-oxopentan-2-yl leucinamide-hydrogen chloride 1/1
CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O)NC(=O)C.Cl
463.02
463
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'