accessibility menu, dialog, popup

UserName

CAS RN 24125-16-4

H3CCH3ONHH3CH3COHNNH2NH2NOHNOCH3HCl

CAS RN 24125-16-4

IUPAC Name: N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide hydrochloride
Synonyms: N-{5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}-2-(2-acetamido-4-methylpentanamido)-4-methylpentanamide hydrochloride
Molecular Weight (g/mol): 463.02
Molecular Formula: C20H39ClN6O4
InChi Key: OQQYPQNFLLAILR-UHFFFAOYNA-N
SMILES: Cl.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)