missing translation for 'onlineSavingsMsg'
Learn More

Leucomalachite Green-d6, MilliporeSigma™ Supelco™

Catalog No. 111018308
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018308 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018308 Supplier Merck Emd Millipore Supplier No. 3394410MG
Only null left
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 1173021-13-0
Physical Form Neat
Linear Formula C6D5 CD[C6H4 N(CH3)2]2
MDL Number MFCD09753325
Synonym Bis-(4-dimethylaminophenyl)phenyl-d5-methane-d1
Type Analytical Standard
Quantity 10 mg
Formula Weight 336.50
Grade Analytical Standard