missing translation for 'onlineSavingsMsg'
Learn More

Leucomalachite Green, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012918
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012918 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012918 Supplier Merck Emd Millipore Supplier No. 6966925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Leucomalachite green is a triphenylmethane dye and a metabolite of malachite green.

Specifications

CAS 129-73-7
Physical Form Neat
Linear Formula C6H5 CH[C6H4 N(CH3)2]2
MDL Number MFCD00008315
Melting Point 100°C to 102°C
Synonym 4,4′-Benzylidenebis(N,N-dimethylaniline)
Type Analytical Standard
Quantity 25 mg
Formula Weight 330.47
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less