missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leucocrystal Violet-d6, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 1173023-92-1 |
| Physical Form | Neat |
| Linear Formula | C25D6H25N3 |
| MDL Number | MFCD11975211 |
| Synonym | Bis(4-dimethylaminophenyl)-4-dimethylamino-d6-phenylmethane |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 379.57 |
| Grade | Analytical Standard |