missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leuco Crystal Violet 98.0+%, TCI America™
Supplier: TCI America M0344250G
Specifications
| Leuco Crystal Violet | |
| 178°C | |
| C25H31N3 | |
| 250 g | |
| OAZWDJGLIYNYMU-UHFFFAOYSA-N | |
| 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline | |
| 69048 | |
| ≥98.0% (HPLC,T) |
| 603-48-5 | |
| Green-White | |
| MFCD00008314 | |
| leucocrystal violet, leuco crystal violet, leucomethyl green, leucogentian violet, 4,4',4-methylidynetris n,n-dimethylaniline, c.i. basic violet 3, leuco, crystal violet leucobase, tris p-dimethylaminophenyl methane, benzenamine, 4,4',4-methylidynetris n,n-dimethyl, 4,4',4-hexamethyltriamino triphenylmethane | |
| CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C | |
| 373.54 | |
| 373.54 | |
| Crystalline Powder |
Chemical Identifiers
| 603-48-5 | |
| 373.54 | |
| OAZWDJGLIYNYMU-UHFFFAOYSA-N | |
| 69048 | |
| CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C |
| C25H31N3 | |
| MFCD00008314 | |
| leucocrystal violet, leuco crystal violet, leucomethyl green, leucogentian violet, 4,4',4-methylidynetris n,n-dimethylaniline, c.i. basic violet 3, leuco, crystal violet leucobase, tris p-dimethylaminophenyl methane, benzenamine, 4,4',4-methylidynetris n,n-dimethyl, 4,4',4-hexamethyltriamino triphenylmethane | |
| 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline |
Safety and Handling
EINECSNumber : (4)-1476
TSCA : Yes