missing translation for 'onlineSavingsMsg'
Learn More

Leuco Crystal Violet 98.0+%, TCI America™

Catalog No. M034425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M034425G 25 g
M0344250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M034425G Supplier TCI America Supplier No. M034425G
Only null left

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym leucocrystal violet, leuco crystal violet, leucomethyl green, leucogentian violet, 4,4',4-methylidynetris n,n-dimethylaniline, c.i. basic violet 3, leuco, crystal violet leucobase, tris p-dimethylaminophenyl methane, benzenamine, 4,4',4-methylidynetris n,n-dimethyl, 4,4',4-hexamethyltriamino triphenylmethane
PubChem CID 69048
IUPAC Name 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 178°C
Color Green-White
Quantity 25 g
Formula Weight 373.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Leuco Crystal Violet
EINECSNumber (4)-1476
TSCA Yes