missing translation for 'onlineSavingsMsg'
Learn More

Leucine, USP, 98.5-101.5%, Spectrum™ Chemical

L1202, 61-90-5, C6H13NO2

Supplier:  Spectrum Chemical Mfg Cor L1202250GM

Quantity 250 g
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18603784


Only null left

Description

Description

Spectrum™ Chemical Leucine, USP is a hydrophobic essential amino acid that is a component in astacin, ferritin and other buffer proteins. It is used for the creation of sterols in the muscle and adipose tissues. Widely used in sport supplements for its muscle building properties, it is also used as a food additive and flavor enhancer. Spectrum Chemical manufactured USP LEUCINE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

61-90-5
131.18
ROHFNLRQFUQHCH-MLHKIVSYNA-N
CC(C)C[C@H](N)C(O)=O
C6H13NO2
MFCD00002617
(2S)-2-amino-4-methylpentanoic acid

Specifications

61-90-5
0.05%
0.2%
Poly Bottle
+14.9° to +17.3°
ROHFNLRQFUQHCH-MLHKIVSYNA-N
(2S)-2-amino-4-methylpentanoic acid
250 g
USP
100%
0.4%
C6H13NO2
MFCD00002617
0.03%
CC(C)C[C@H](N)C(O)=O
131.18
98.5 to 101.5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions