missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Leucine (L), USP
Description
Leucine (L), USP come as white crystals, with a characteristic odor.
Validation of grade in progress.
Alternative name(s): L-Leucine
Specifications
Specifications
| Chemical Name or Material | Leucine (L), USP |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 61-90-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 131.17 |
| Quantity | 100 g |
| Percent Purity | 98.5 to 101.5 % |
| Grade | USP |