missing translation for 'onlineSavingsMsg'
Learn More

Letaplimab - Research Grade Biosimilar, ProSci

Catalog No. 16834580
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16834580 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16834580 Supplier ProSci Llc Supplier No. 219250100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chinese Hamster (CHO) Recombinant Monoclonal Antibody

Letaplimab Recombinant Monoclonal Antibody specifically detects Letaplimab.

Specifications

Antigen Letaplimab
Applications Not Available
Classification Recombinant Monoclonal
Conjugate Unconjugated
Formulation PBS buffer pH 7.5
Gene Accession No. Q08722
Gene Alias HUMAN ANTI-CD47 MONOCLONAL ANTIBODY (IBI-188), IBI188, IBI-188, IMMUNOGLOBULIN G4 (223-PROLINE,229-ALANINE,230-ALANINE,DE-C-TERMINAL-LYSINE), ANTI-(HUMAN CD47 ANTIGEN) (HUMAN MONOCLONAL IBI188 .GAMMA.4-CHAIN), DISULFIDE WITH HUMAN MONOCLONAL IBI188 .KAPPA.-CHAIN, DIMER LETAPLIMAB, LETAPLIMAB [INN], IAP; OA3; MER6
Host Species Chinese Hamster (CHO)
Purification Method >95%
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 961
Target Species Not Available
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Store at +4°C short term (1-2 weeks). Store at -20 °C 12 months. Store at -80°C long term.
Product Type Antibody
Form Purified
Isotype IgG4 κ
Research Category Other
Show More Show Less