Learn More
Invitrogen™ Human Leptin Instant ELISA™ Kit
Description
The Human Leptin) ELISA quantitates Hu Leptinin human serum, plasma or other body fluids. The assay will exclusively recognize both natural and recombinant Hu Leptin. Principle of the method The Human Leptin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P41159 |
| Assay Range | 63-4000 pg/mL |
| Assay Sensitivity | 20.0 pg/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human Leptin, Biotin-Conjugate (anti-human Leptin monoclonal antibody), Streptavidin-HRP and Assay Buffer, lyophilized Aluminum pouch(es) with a human Leptin Standard curve (coloured) Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20) Assay Buffer Concentrate 20x (Use when an external predilution of the samples is needed) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Adhesive Films |
| Conjugate | HRP |
| Immunoassay Kit Format | Coated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |