missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lepidine 97.0+%, TCI America™
$223.33 - $2477.69
Chemical Identifiers
| CAS | 491-35-0 |
|---|---|
| Molecular Formula | C10H9N |
| Molecular Weight (g/mol) | 143.189 |
| MDL Number | MFCD00006784 |
| InChI Key | MUDSDYNRBDKLGK-UHFFFAOYSA-N |
| Synonym | lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 |
| PubChem CID | 10285 |
| ChEBI | CHEBI:48983 |
| IUPAC Name | 4-methylquinoline |
| SMILES | CC1=CC=NC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L002425ML
|
TCI America
L002425ML |
25 mL |
Each for $223.33
|
|
|||||
|
L0024100ML
|
TCI America
L0024100ML |
100 mL |
Each for $619.90
|
|
|||||
|
L0024500ML
|
TCI America
L0024500ML |
500 mL |
Each for $2,477.69
|
|
|||||
Chemical Identifiers
| 491-35-0 | |
| 143.189 | |
| MUDSDYNRBDKLGK-UHFFFAOYSA-N | |
| 10285 | |
| 4-methylquinoline |
| C10H9N | |
| MFCD00006784 | |
| lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 | |
| CHEBI:48983 | |
| CC1=CC=NC2=CC=CC=C12 |
Specifications
| 491-35-0 | |
| Yellow | |
| C10H9N | |
| 25 mL | |
| MUDSDYNRBDKLGK-UHFFFAOYSA-N | |
| 4-methylquinoline | |
| 10285 | |
| 143.19 | |
| Liquid |
| 10°C | |
| 265°C | |
| MFCD00006784 | |
| lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894 | |
| CC1=CC=NC2=CC=CC=C12 | |
| 143.189 | |
| CHEBI:48983 | |
| ≥97.0% (GC,T) | |
| Lepidine |
Safety and Handling
EINECSNumber : (5)-0796
RTECSNumber : OH0316000
TSCA : Yes