missing translation for 'onlineSavingsMsg'
Learn More

Lepidine 97.0+%, TCI America™

Supplier:  TCI America L0024100ML

 View more versions of this product

Catalog No. L0024100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lepidine
10°C
265°C
MFCD00006784
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
CC1=CC=NC2=CC=CC=C12
143.189
CHEBI:48983
≥97.0% (GC,T)
491-35-0
Yellow
C10H9N
100 mL
MUDSDYNRBDKLGK-UHFFFAOYSA-N
4-methylquinoline
10285
143.19
Liquid

Chemical Identifiers

491-35-0
143.189
MUDSDYNRBDKLGK-UHFFFAOYSA-N
10285
4-methylquinoline
C10H9N
MFCD00006784
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
CHEBI:48983
CC1=CC=NC2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0796

RTECSNumber : OH0316000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions