missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ Cas9 Lentivirus

Catalog No. A32069
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32069 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. A32069 Supplier Invitrogen™ Supplier No. A32069

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Research Use Only. Not for use in diagnostic procedures.

LentiArray Cas9 Lentivirus is provided as high-titer purified lentiviral particles and is an efficient method to drive high level Cas9 nuclease expression in a wide variety of cell types.

The LentiArray Cas9 Lentivirus construct includes a human codon-optimized version of Cas9 with two nuclear localization signals to facilitate efficient delivery into the nucleus. The 1-mL size is designed to support studies utilizing a co-infection approach (Cas9 lentivirus + guideRNA lentivirus delivered together) with either the LentiArray CRISPR libraries or LentiArray gRNAs against single gene targets. LentiArray Cas9 Lentivirus is provided as ready-to-use lentivirus at a titer of 1 x 107 TU/mL.

Features include:

  • 1 mL high-titer lentiviral particles
  • Ready-to-use lentiviral particles at a titer of 1 x 107 TU/mL
  • EF-1a promotor that is highly effective in a wide variety of cell models
  • Create stable Cas9-expressing cell models for screening and focused CRISPR knockout studies

Recommended Storage

Store at -68 to -85°C

Specifications

Content And Storage 1 x 1 mL. Store at -68 to -85°C.
Delivery Type Lentiviral
Sample Type siRNA
Promoter EF-1a
Transfection Technique Lentiviral transduction
Species Human
For Use With (Application) CRISPR gene knockout
Product Line LentiArray
Product Type Cas9 Lentivirus
Quantity 1 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.