missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lectin, immobilized on Agarose CL-4B from Triticum vulgaris for affinity chromatography, MilliporeSigma™ Supelco™
$3741.02 - $3741.02
Specifications
| Synonym | Wheat germ agglutinin, immobilized on agarose CL-4 B |
|---|---|
| Type | Organic Standard |
| Grade | Affinity Chromatography |
Specifications
| Wheat germ agglutinin, immobilized on agarose CL-4 B | |
| Organic Standard | |
| Affinity Chromatography |