missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LECT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150275
Description
LECT2 Polyclonal Antibody for IHC (P)
This gene encodes a secreted, 16 kDa protein that acts as a chemotactic factor to neutrophils and stimulates the growth of chondrocytes and osteoblasts. This protein has high sequence similarity to the chondromodulin repeat regions of the chicken myb-induced myeloid 1 protein. A polymorphism in this gene may be associated with rheumatoid arthritis.Specifications
| LECT2 | |
| Polyclonal | |
| Unconjugated | |
| LECT2 | |
| chm2; ChM-II; chondromodulin II; chondromodulin-II; hLECT2; Lect2; LECT-2; leukocyte cell derived chemotaxin 2; leukocyte cell-derived chemotaxin 2; leukocyte cell-derived chemotaxin-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O14960 | |
| LECT2 | |
| Recombinant Leukocyte Cell Derived Chemotaxin 2 (LECT2) (Gly19-Leu151). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |