missing translation for 'onlineSavingsMsg'
Learn More

Medisca Lecithin (Granular)

Catalog No. 14186058
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186058 2.5 kg
14185586 1 kg
14185490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186058 Supplier Medisca Supplier No. 094701

Lecithin comes as golden to light tan granules with a neutral odor. It is hygroscopic.

Lecithin comes as golden to light tan granules with a neutral odor. It is hygroscopic.

Specifications

Chemical Name or Material Lecithin (Granular)
Packaging Pail, 8 Liter, White, HDPE, Square
CAS 8002-43-5
Type Drug Standard
Molecular Weight (g/mol) Not applicable
Quantity 2.5 kg
Grade Ungraded