Learn More
New Pig Leak and Drip Pad System
Keep oily leaks and drips off the floor with this refillable holder and absorbent mat pad system
Supplier:  New Pig PAD202
Catalog No. 17-987-840
Only null left
Explore available promotions
Description
Description
  • Refillable system for cost savings: This system allows you to replace only the absorbent mat, reducing waste and the long-term cost of managing leaks and drips.
  • Keeps floors clean and safe: The absorbent mat pad sits within a durable holder, effectively capturing oily leaks and drips before they can reach the floor and create a slip hazard.
  • Versatile absorbent options: The system can be used with a variety of PIG Absorbent Mats, allowing you to choose the right pad for your specific fluid and absorbency needs.
  • Durable and easy to clean: The rigid, plastic holder is built to withstand industrial environments and is easy to wipe down and clean.
  • Low-profile design: The system's low profile allows it to be placed directly under equipment or machinery without getting in the way.
  • Reduces cleanup time: By containing drips at their source, this system saves valuable time and effort that would otherwise be spent on cleaning up messier spills.
Specifications
Specifications
Oil Only Leak and Drip Pad
Product Suggestions
Product Suggestions