missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Leadmium™ Green AM dye
Description
Leadmium™ Green AM dye
The proprietary Leadmium™ Green AM dye from Molecular Probes™ provides researchers with a unique tool to specifically measure intracellular lead and cadmium levels with high sensitivity. While standards assays exist for measuring lead and cadmium in solution, the unique Leadmium™ Green AM indicator was specifically designed to detect these toxic environmental contaminants inside living cells.
- Highly specific indicator for intracellular measurements of lead and cadmium
- Flexible format allows use on multiple formats - flow cytometry, fluorescence microscopy imaging and plate readers
- Bright fluorescence and high discrimination from calcium allows detection of nanomolar levels of lead and micromolar levels of cadmium
- Convenient excitation with 488 nm laser line
- The Leadmium™ Green AM dye is readily loaded and is retained very well following esterase cleavage of the acetoxymethyl group
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Ion Indicator |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Dye Type | Leadmium Green |
| Product Line | Leadmium |
| Quantity | 5 x 50 μg |
| Shipping Condition | Room Temperature |
| Excitation Wavelength | 490 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.