missing translation for 'onlineSavingsMsg'
Learn More

Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals

Used in piezoelectric devices and electro-optic devices

$175.64 - $656.70

Chemical Identifiers

CAS 12626-81-2
Molecular Formula O5PbTiZr
Molecular Weight (g/mol) 426.286
MDL Number MFCD00212270
InChI Key HFGPZNIAWCZYJU-UHFFFAOYSA-N
Synonym lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
PubChem CID 159452
IUPAC Name lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3975809
View Documents
Thermo Scientific Chemicals
03975809
10 g N/A
Only null left
 
AA3975818
View Documents
Thermo Scientific Chemicals
03975818
50 g N/A
Only null left
 
Description

Description

Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12626-81-2
426.286
HFGPZNIAWCZYJU-UHFFFAOYSA-N
159452
[O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
O5PbTiZr
MFCD00212270
lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
lead(2+);oxygen(2-);titanium(4+);zirconium(4+)

Specifications

Yellow
Semi-solid, Viscous Liquid
O5PbTiZr
UN1993
Soluble in alcohols
[O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
426.286
Oxide ≈30 wt%
12626-81-2
10 g
MFCD00212270
lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
HFGPZNIAWCZYJU-UHFFFAOYSA-N
lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
159452
Lead zirconium titanium oxide, Polymeric precursor
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H304-H373-H318-H302-H336-H315
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes skin irritation.

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P331-P332+P313-P362-P501c

H302+H332-H304-H315-H319-H336-H350-H360Df-H373

DOTInformation : Hazard Class: 3; Packaging Group: II

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only