CAS RN 12626-81-2
IUPAC Name:
titanium(4+) zirconium(4+) λ²-lead(2+) pentaoxidandiide
Synonyms:
lead zirconate titanate
Molecular Weight (g/mol):
426.3
Molecular Formula:
O5PbTiZr
InChi Key:
HFGPZNIAWCZYJU-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[O--].[Ti+4].[Zr+4].[Pb++]
1
–
1
of
1
results
1
Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals
CAS: 12626-81-2 Molecular Formula: O5PbTiZr Molecular Weight (g/mol): 426.286 MDL Number: MFCD00212270 InChI Key: HFGPZNIAWCZYJU-UHFFFAOYSA-N Synonym: lead zirconate titanate,lead titanate zirconate,lead titanium zirconium oxide,lead zirconium titanate,pzt-c,plumbum zirconate-titanate,pzt 5,pzt 8,lead titanium zirconium oxide pb ti,zr o3,lead zirconium titanium oxide PubChem CID: 159452 IUPAC Name: lead(2+);oxygen(2-);titanium(4+);zirconium(4+) SMILES: [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]