missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals
Description
Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Semi-solid, Viscous Liquid |
| Quantity | 10 g |
| UN Number | UN1993 |
| Solubility Information | Soluble in alcohols |
| Concentration or Composition (by Analyte or Components) | Oxide ≈30 wt% |
| Chemical Name or Material | Lead zirconium titanium oxide, Polymeric precursor |
Safety and Handling
| Hazard Category | H302+H332-H304-H315-H319-H336-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H225-H360-H304-H373-H318-H302-H336-H315 Highly flammable liquid and vapor. May damage fertility or the unborn child. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Causes serious eye damage. Harmful if swallowed. May cause drowsiness or dizziness. Causes skin irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P331-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| TSCA | Yes |
RUO – Research Use Only