missing translation for 'onlineSavingsMsg'
Learn More

Lead Sulfate, Powder, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L109525KG

 View more versions of this product

Catalog No. 18611261


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7446-14-2
2.5 kg
Reagent
1
97%
Amber Glass Bottle

Chemical Identifiers

7446-14-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.