missing translation for 'onlineSavingsMsg'
Learn More

Lead Sulfate, Powder, 97%, Spectrum™ Chemical

Catalog No. p-7165781
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611259 125 g Amber Glass Bottle
18611263 500 g Amber Glass Bottle
18611261 2.5 kg Amber Glass Bottle
18611260 12 kg Poly Pail
18611262 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611259 Supplier Spectrum Chemical Mfg Cor Supplier No. L1095125GM
Only null left

Chemical Identifiers

CAS 7446-14-2

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.