missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, For IC, 1,000 mg/L Pb+2 in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111002661
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002661 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002661 Supplier Merck Emd Millipore Supplier No. 51777100ML

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.

Specifications

Product Line TraceCERT
Concentration 1000 mg/L Pb +2 in. nitric acid
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC