missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, 100 ppm Pb, Ricca Chemical

Catalog No. R4295000100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4295000100 100 mL
429516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R4295000100 Supplier Ricca Chemical Company Supplier No. R4295000100A
Only null left

Specifications

Chemical Name or Material Lead Standard
Packaging Natural Poly Bottle
Name Note 1mL = 0.1mg Pb, 100ppm Pb
CAS 7439-92-1, 7697-37-2
Assay Percent Range 100.0 ± 0.5ppm
Physical Form Liquid
Molecular Formula Pb
MDL Number MFCD00134050
Color Brown
Boiling Point 100°C
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES [Pb]
Solvent or Matrix Water
IUPAC Name lead
Molecular Weight (g/mol) 207.20
Quantity 100 mL
PubChem CID 5352425
ChEBI CHEBI:27889
Show More Show Less