missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Red Powder, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L1085500GM

 View more versions of this product

Catalog No. 18611244


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1314-41-6
500 g
XMFOQHDPRMAJNU-UHFFFAOYSA-N
bicyclo[3.1.1]triplumboxan-1-yl
98%
Amber Glass Bottle
1
O4Pb3
O1[Pb]2O[Pb]1O[Pb]O2
685.60
Reagent

Chemical Identifiers

1314-41-6
685.60
bicyclo[3.1.1]triplumboxan-1-yl
O4Pb3
XMFOQHDPRMAJNU-UHFFFAOYSA-N
O1[Pb]2O[Pb]1O[Pb]O2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.