missing translation for 'onlineSavingsMsg'
Learn More

Lead Oxide, Red Powder, 98%, Spectrum™ Chemical

Catalog No. p-7165778
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18611242 125 g Amber Glass Bottle
18611244 500 g Amber Glass Bottle
18611245 2.5 kg Amber Glass Bottle
18611243 12 kg Poly Pail
18611246 45 kg Fiber Drum
5 options

Catalog No. 18611242

Supplier: Spectrum Chemical Mfg Cor L1085125GM

Only null left

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.60
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
IUPAC Name bicyclo[3.1.1]triplumboxan-1-yl
SMILES O1[Pb]2O[Pb]1O[Pb]O2

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.