missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals

$298.34 - $298.34

Chemical Identifiers

CAS 4146-73-0
Molecular Formula C4F6O4Pb
Molecular Weight (g/mol) 433.23
MDL Number MFCD00061594
InChI Key XVULNTYAIWEMSW-UHFFFAOYSA-L
Synonym lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
PubChem CID 71311182
IUPAC Name lead(2+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1755309
View Documents
Thermo Scientific Chemicals
01755309
10 g
Each for $298.34
Only null left
 
Description

Description

It is employed in pyrolytic deposition of fluoride glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4146-73-0
433.23
XVULNTYAIWEMSW-UHFFFAOYSA-L
71311182
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]
C4F6O4Pb
MFCD00061594
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
lead(2+);2,2,2-trifluoroacetate

Specifications

4146-73-0
White
C4F6O4Pb
MFCD00061594
UN2291
Soluble in water,trifluoroacetic acid,tetrahydrofuran,acetone.
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]
433.23
442.23(433.22 Anhydrous)
Lead(II) trifluoroacetate hemihydrate
147°C to 153°C
Odorless
Pb(CF3COO)2·0.5H2O
10 g
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
XVULNTYAIWEMSW-UHFFFAOYSA-L
lead(2+);2,2,2-trifluoroacetate
71311182
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only