missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™
$107.80 - $327.28
Chemical Identifiers
| CAS | 15187-16-3 |
|---|---|
| Molecular Formula | C32H16N8Pb |
| MDL Number | MFCD00053951 |
| Synonym | Phthalocyanine Lead(II) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L0230200MG
|
TCI America
L0230200MG |
200 mg |
Each for $107.80
|
|
|||||
|
L02301G
|
TCI America
L02301G |
1 g |
Each for $327.28
|
|
|||||
Chemical Identifiers
| 15187-16-3 | |
| C32H16N8Pb | |
| MFCD00053951 | |
| Phthalocyanine Lead(II) |
Specifications
| 15187-16-3 | |
| C32H16N8Pb | |
| 200 mg | |
| Phthalocyanine Lead(II) | |
| ≥98.0% (T) | |
| Lead(II) Phthalocyanine (purified by sublimation) |
| Blue-Purple | |
| MFCD00053951 | |
| 2291 | |
| 719.74 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes