missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™

Supplier:  TCI America L02301G

 View more versions of this product

Catalog No. L02301G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lead(II) Phthalocyanine (purified by sublimation)
Blue-Purple
MFCD00053951
2291
719.74
Crystalline Powder
15187-16-3
C32H16N8Pb
1 g
Phthalocyanine Lead(II)
≥98.0% (T)

Chemical Identifiers

15187-16-3
C32H16N8Pb
MFCD00053951
Phthalocyanine Lead(II)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions