missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Nitrate, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥99.0%

$975.40 - $975.40

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name lead(2+);dinitrate
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000894
View Documents
Honeywell-Fluka
22862125KG
2.5 kg
Each for $975.40
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10099-74-8
331.20
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
lead(2+);dinitrate
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
CHEBI:37187

Specifications

10099-74-8
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
331.20
CHEBI:37187
≥99%
Lead(Ii) Nitrate
2.5 kg
Pb(NO3)2
1469
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead(2+);dinitrate
24924
331.21g/mol
ACS Reagent
Videos
Safety and Handling

Safety and Handling

P201-P210-P220-P280-P305 + P351 + P338 + P310-P308 + P313

H272-H302 + H332-H318-H360Df-H373-H410

EINECSNumber : 233-245-9

RTECSNumber : OG2100000

SDS
missing translation for 'documents'

missing translation for 'documents'