missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) fluoride, Puratronic™, 99.997% (metals basis)

Catalog No. p-7048514
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AA1072314 25 g
AA1072322 100 g
2 options

Catalog No. AA1072314

Supplier: Thermo Scientific Chemicals 01072314

Only null left

CAS: 7783-46-2 | F2Pb | 245.20 g/mol

Lead(II) fluoride is used in fuses, glass coatings to reflect infrared rays and phosphors for television screens and in low melting glasses. Further, it serves as a catalyst for the preparation of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-46-2
Molecular Formula F2Pb
Molecular Weight (g/mol) 245.20
MDL Number MFCD00011162
InChI Key FPHIOHCCQGUGKU-UHFFFAOYSA-L
Synonym lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
PubChem CID 24549
IUPAC Name difluorolead
SMILES [F-].[F-].[Pb++]

Specifications

Boiling Point 1290°C
Melting Point 855°C
Physical Form Powder
Quantity 25 g
UN Number UN2291
Merck Index 14,5408
Solubility Information Soluble in nitric acid. Slightly soluble in water. Insoluble in acetone and ammonia.
Formula Weight 245.2
Percent Purity 99.997%
Grade Puratronic
Odor Odorless
Density 8.445 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) fluoride
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 231-998-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only