accessibility menu, dialog, popup

UserName

FFPb

CAS RN 7783-46-2

IUPAC Name: λ²-lead(2+) difluoride
Synonyms: lead(II) fluoride
Molecular Weight (g/mol): 245.2
Molecular Formula: F2Pb
InChi Key: FPHIOHCCQGUGKU-UHFFFAOYSA-L
SMILES: [F-].[F-].[Pb++]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)