missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) fluoride, 99% min

Catalog No. p-7080448
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8828230 250 g
AA88282A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8828230 Supplier Thermo Scientific Chemicals Supplier No. 08828230

CAS: 7783-46-2 | F2Pb | 245.20 g/mol

Lead(II) fluoride is a specialty product for proteomics research. It is used in low melting glasses, in glass coatings to reflect infrared rays, in phosphors for television-tube screens and as a catalyst for the manufacture of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-46-2
Molecular Formula F2Pb
Molecular Weight (g/mol) 245.20
MDL Number MFCD00011162
InChI Key FPHIOHCCQGUGKU-UHFFFAOYSA-L
Synonym lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
PubChem CID 24549
IUPAC Name difluorolead
SMILES [F-].[F-].[Pb++]

Specifications

Boiling Point 1290°C
Melting Point 855°C
Physical Form Powder
Quantity 250 g
UN Number UN2291
Merck Index 14,5408
Solubility Information Sparingly soluble in water(0.64g/L at 20°C). Soluble in nitric acid. Insoluble in acetone and ammonia.
Formula Weight 245.2
Percent Purity ≥99%
Odor Odorless
Density 8.445 g/mL
Chemical Name or Material Lead (II) fluoride
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 231-998-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only