missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis)

Catalog No. p-7048511
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1071906 5 g
AA1071918 50 g
AA1071930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1071906 Supplier Thermo Scientific Chemicals Supplier No. 01071906
Only null left

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol

Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75°C
Physical Form Crystalline
Quantity 5 g
UN Number UN1616
Beilstein 3730298
Merck Index 14,5397
Solubility Information Freely soluble in water,alcohol and glycerol.
Formula Weight 379.33 (325.28 Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Density 2.55 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) acetate trihydrate
Show More Show Less
Hazard Category H360D-H361f-H372
Hazard Statement GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber 206-104-4
RTECSNumber OF8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only