Learn More
Lead(II) acetate trihydrate, 99+%, ACS reagent
CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Supplier: Thermo Scientific Chemicals 423841000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lead(II) acetate trihydrate | |
| 6080-56-4 | |
| Crystalline Powder and/or Crystals | |
| 99.0 to 103% (Complexometry) | |
| (CH3CO2)2Pb·3H2O | |
| 01,532; 02,233; 04,276 | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| λ²-lead(2+) diacetate trihydrate | |
| 22456 | |
| 379.33 | |
| ACS Reagent | |
| Decomposes information: >100°C |
| 75.0°C | |
| White | |
| 100 g | |
| C4H12O7Pb | |
| MFCD00150023 | |
| 15, 5452 | |
| Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O | |
| 379.30 | |
| CHEBI:33112 | |
| ≥99% | |
| Plastic bottle | |
| 13.1 |
Chemical Identifiers
| 6080-56-4 | |
| 379.30 | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| 22456 | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
| C4H12O7Pb | |
| MFCD00150023 | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| CHEBI:33112 |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA
GHS Signal Word: Danger
RTECSNumber : OF8050000
RUO – Research Use Only